Interceptor® Plus Level A, Gas-Tight and Vapor-Tight Chemical Protective Encapsulated Suit with Zip Rear Entry and Expanded Back for SCBA, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Level A, chemcial protective gas-tight and vapor-tight encapsulated suits are made with Interceptor® Plus chemcial protective fabric for proven protection against a broad spectrum of toxic industrial chemicals, ammonia, and fentanyl.
- Interceptor® Plus chemical protective film
- Level A encapsulated suit
- Gas-tight and vapor-tight
- Rear entry design
- Expanded back to accommodate SCBA
- Supported by chemical toxicity risk modeling software
Interceptor® Plus chemical protectve level A encapsulated suits provide protection from the most toxic industrial chemicals and hazardous materials. These garments feature a rear entry design with an expanded back to accommodate an SCBA.
Specifications
- Catalog No:
- 76314-202
- 76314-204
- 76314-206
- 76314-208
- 76314-210
- 76314-212
- Seam type:
- Heat sealed plus
- Heat sealed plus
- Heat sealed plus
- Heat sealed plus
- Heat sealed plus
- Heat sealed plus
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection