About this item
Stabilize DNA at ambient temperature for transport and storage.
GenTegra DNA protects DNA from hydrolysis and oxidation, stabilizing it for long-term ambient temperature storage, providing better protection from freezing which only slows those processes.
DNA samples stored in GenTegra DNA show no degradation after the equivalent of 20 years of storage at ambient temperature (via accelerated aging studies).
Simple rehydration process for 100% sample recovery for downstream applications, such as transfections, quantitative PCR, gene expression, next-generation sequencing, genotyping, HLA typing, forensics, RRN profiling.