All models feature a unique snap-valve pour spout that is easily popped open to provide dripless pouring.
- Unique snap-valve pour spout is easily popped open
- Dripless pouring
- Specially designed leakproof screw cap with liner for safer transport
- Metal-free and resistant to hydrochloric acid
- Large handle can be gripped comfortably
- Volume level read in 50 ml increments
- Can be gamma sterilized
This more conventionally styled urine bottle is available in 2.5 and 3.5 l sizes. It also incorporates some of the great features of the URISAFE® 24 Hour Urine Collection Container. The innovative cap has a leakproof design with a cap liner to ensure safe sample transport. The unique snap-valve pour allows spout to easily pop open and push shut. Also offers a controlled flow rate for better handling of poured volumes with reduced risks of aerosol contamination when pouring.
Pour spout eliminates splashing and exposure to hazardous body fluids. A large central handle can be gripped comfortably to reduce fatigue, and permits pouring off of samples with ease.
For IVD use. Chemically resistant. Metal- and latex-free. Zinc- and fluorescence-free. Container can be subjected to freezing, thawing, and EtO gas sterilization without causing changes in materials or physical appearance.