PermaLife™ cell culture bags represent the optimal way to harvest, expand, concentrate and freeze critical cell cultures.
- Contains no plasticizers or chemical residuals
- Possibility of contamination is less
- Suitable for cryogenic preservation
- Temperature resistant from –200 to 200 °C
- Optical transmission of greater than 95%
Made of FEP, it is inert, so antibodies, cytokines and other genetic material are unaffected by these bags. They are gas permeable and provide a high rate of oxygen transfer to cells under culture. Water permeability is very low, so water loss is negligible and humidified incubators are not required.
Swabable, needle-free valves are provided on every bag to preserve sterile access and reduce the chance of needle sticks. No significant decrease in mechanical strength at temperatures from cryogenic storage (–196 °C) to high autoclave (137 °C).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- MSPP-PL30
- MSPP-PL120
- MSPP-PL325
- MSPP-PL240
- MSPP-PL70
- O₂ (gas) cc/day @ 37 °C:
- 126 at 25 °C
- 288 at 25 °C
- 842 at 25 °C
- 627 at 25 °C
- 199 at 25 °C
- CO₂:
- 281 cc/day at 25 °C
- 643 cc/day at 25 °C
- 1879 cc/day at 25 °C
- 1400 cc/day at 25 °C
- 444 cc/day at 25 °C
- N2 (gas) cc/day @ 37 °C:
- 54 at 25 °C
- 124 at 25 °C
- 363 at 25 °C
- 270 at 25 °C
- 86 at 25 °C
- H₂O (Liquid) cc/day @ 37 °C:
- 0.08 at 25 °C
- 0.17 at 25 °C
- 0.51 at 25 °C
- 0.38 at 25 °C
- 0.12 at 25 °C