Order Entry
United States
ContactUsLinkComponent
Recombinant Red Fluorescent Protein (RFP) Control
Recombinant Red Fluorescent Protein (RFP) Control
Catalog # RL000-001-379
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Recombinant Red Fluorescent Protein (RFP) Control
Catalog # RL000-001-379
Supplier Number:  000-001-379
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein Synonyms:
    DsRed
  • Protein/Peptide Name:
    Red Fluorescent Protein (RFP) Control
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    RL000-001-379
  • Supplier no.:
    000-001-379

Specifications

About this item

Recombinant Red Fluorescent Control Protein (RFP) Control

  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot

Recombinant RFP (wild type, sequence corresponding to RFP, Discosoma spp.) is intended for use as a control when using polyclonal or monoclonal anti-RFP in immunological assays, in fluorometry, and in fluorescence activated cell sorting (FACS). Polyclonal anti-RFP can detect RFP by ELISA and western blot. Biotin conjugated polyclonal anti-RFP used in a sandwich ELISA is well suited to titrate RFP in solution when used in combination with unconjugated anti-RFP antibody. This product can be used as a control in western blots.

RFP is suitable as a control for polyclonal or monoclonal anti-RFP in immunological assays. The molecular weight of RFP (calculated) is 27 kDa, but in a gel it is expected to run at approximately 28-30 kDa.