Order Entry
United States
ContactUsLinkComponent
Green Fluorescent Protein (GFP) Control Protein
Green Fluorescent Protein (GFP) Control Protein
Catalog # RL000-001-215
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Green Fluorescent Protein (GFP) Control Protein
Catalog # RL000-001-215
Supplier Number:  000-001-215
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein/Peptide Name:
    Green Fluorescent Protein (GFP) Control Protein
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    Control Protein
  • Cat. No.:
    RL000-001-215
  • Supplier no.:
    000-001-215

Specifications

About this item

Green Fluorescent Protein (GFP) Control Protein

  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot

Green Fluorescent Protein (GFP) is used as a positive control when detecting GFP fusion proteins in anti-GFP antibody assays. Polyclonal anti-GFP can detect GFP by ELISA (sandwich or capture) or western blot. Biotin conjugated polyclonal anti-GFP used in a sandwich ELISA is well suited to titrate GFP in solution when used in combination with monoclonal anti-GFP (p/n 600-301-215), using either form of the antibody as the capture or detection antibody. Control GFP can be used as a control in western blots. Researchers should determine optimal titers for applications.

GFP is suitable as a control for polyclonal or monoclonal anti-GFP in immunological assays. The molecular weight of GFP is 28 kDa.