Trio.Bas™ MONO Kits with Data Transfer Cable, Hardy Diagnostics
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
- Aspirating head with easy manipulation bayonet closure
- Manual or automatic operations
- Ergonomic and balanced design to facilitate handling with ease
The Trio.Bas™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area.
All air samplers are lightweight, with an antibacterial techno-polymer shockproof body, and are IP65 protected from dust and water. Per USP <797>, Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.
: ISO 14698, USP 797, USP 116, EU GPL-GMP, ISO 17025.
: Additional accessory components are available separately.
: Trio.Bas™ MONO Kit Contains:
Trio.Bas™ MONO, Contact Plate format, includes calibration certificate
Transfer Cable for transferring data from instrument directly to PC
Standard wall charger; Simple AC power cord connects to air sampler unit
Stainless steel aspirating head, autoclavable
Stainless steel cover head, autoclavable
Robust hard-shell carrying case to protect the sampler in transit
IQ, OQ, PQ, fillable documents
Trio.Bas™ MONO, Contact Plate format, includes calibration certificate
Transfer Cable for transferring data from instrument directly to PC
Standard wall charger; Simple AC power cord connects to air sampler unit
Stainless steel aspirating head, autoclavable
Stainless steel cover head, autoclavable
Robust hard-shell carrying case to protect the sampler in transit
IQ, OQ, PQ, fillable documents