Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant IL-6 R alpha (from HEK293 Cells)
Catalog # MSPP-8954SR025
Supplier:  R&D Systems
CAS Number:  
Human Recombinant IL-6 R alpha (from HEK293 Cells)
Catalog # MSPP-8954SR025
Supplier:  R&D Systems
Supplier Number:  8954-SR-025
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    25 μg
  • Environmentally Preferable:
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    3570
  • Protein Synonyms:
    interleukin 6 receptor|IL-6RA|MGI-2A|IL6Q|IL6RA|BSF-2|IL6R alpha|IL6RQ|IL6|IL-6R-1|IL6R|CD126|gp80
  • Accession No.:
    P08887 (IL-6 R alpha) & P05231 (IL-6)
  • Protein/Peptide Name:
    IL-6 R alpha
  • Purity:
    >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
  • Molecular Weight:
    78-93 kDa, reducing conditions
  • Cat. No.:
    MSPP-8954SR025

Specifications

About this item

The Recombinant Human IL-6/IL-6 R alpha Protein Chimera from R&D Systems is derived from HEK293. The Recombinant Human IL-6/IL-6 R alpha Protein Chimera has been validated for the following applications: Bioactivity.