To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:3084
- Protein Synonyms:Neuregulin1 beta 1|HRG1|NDF|Heregulin-1|SMDF|MSTP131|ARIA|NRG1-IT2|HRG|NRG1|GGF2|HRG1-beta 1|neuregulin 1|MST131|GGF|Neuregulin1|HRG1-alpha|HRGA|HGL
- Accession No.:AAA58639
- Protein/Peptide Name:Neuregulin-1 beta 1/NRG1 beta 1
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Cat. No.:220024-402
- Supplier no.:377-HB-050
The Recombinant Human NRG1-beta 1/HRG1-beta 1 ECD Protein from R&D Systems is derived from E. coli. The Recombinant Human NRG1-beta 1/HRG1-beta 1 ECD Protein has been validated for the following applications: Bioactivity.