MBX-8025 ≥98% (by HPLC)
Supplier: Avantor
Certificates
About this item
Synonyms: Seladelpar, (R)-2-(4-((2-ethoxy-3-(4-(trifluoromethyl)phenoxy)propyl)thio)-2-methylphenoxy)acetic acid, RWJ-800025
CAS Number:851528-79-5
:
Formula:C₂₁H₂₃F₃O₅S
MDL Number:MFCD28502443
Molecular Weight:444.46 g/mol
Specifications
- Environmentally Preferable: