SharpsMaster HV® 7082 Gloves, HexArmor
Supplier: Avantor
Certificates
About this item
The SharpsMaster HV® 7082 is the high visibility solution that utilizes SuperFabric® brand material technology engineered to provide the highest levels of needle and cut protection available on the market.
- Highest needlestick resistance available provided by the layering of SuperFabric® brand material
- Tested using actual 25 gauge needles against ANSI/ISEA 105 standard
- Highest level of cut resistance on noted enhanced areas, ANSI/ISEA Cut Level A9 and Needlestick Level 4
- Flat nitrile three-quarter knuckle dip
- Launderable
This glove gives end users incredible protection in a single glove solution. Don't get stuck with faulty hand protection - become a Sharpsmaster.