Order Entry
United States
ContactUsLinkComponent
SP6 Promoter Primer, 2 µg, Promega
SP6 Promoter Primer, 2 µg, Promega
Catalog # PAQ5011
CAS Number:  
undefined
SP6 Promoter Primer, 2 µg, Promega
Catalog # PAQ5011
Supplier Number:  Q5011
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    SP6 Promoter Primer, 2microg
  • Size:
    2 μg
  • Environmentally Preferable:
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAQ5011
  • Supplier no.:
    Q5011

Specifications

About this item

The SP6 Promoter Sequencing Primer is used for sequencing inserts cloned into various vectors.

  • Designed for Sequencing Inserts Cloned into the pGEM(R), pALTER(R)-MAX and pCI-neo Vectors
  • Designed to be annealed to single-stranded DNA or, after alkaline denaturation, to double-stranded DNA
  • Purified by gel electrophoresis or HPLC
  • Primer Sequence: 5´-d(TATTTAGGTGACACTATAG)-3´
  • Supplied at a concentration of 10µg/ml

The SP6 Promoter Primer is designed for sequencing inserts cloned into the pGEM, pALTER-MAX and pCI-neo Vectors. The primer is designed to be annealed to single-stranded DNA or, after alkaline denaturation, to double-stranded DNA. The promoter primer is purified by gel electrophoresis or HPLC. Primer Sequence: 5'-d(TATTTAGGTGACACTATAG)-3'.