ContactUsLinkComponent
Connectors, Luer, BAXTER IV SOLUTIONS
Connectors, Luer, BAXTER IV SOLUTIONS
Catalog # 10847-610
Supplier:  Baxter
Connectors, Luer, BAXTER IV SOLUTIONS
Catalog # 10847-610
Supplier:  Baxter
Supplier Number:  H93813701
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    RAPIDFILL connectors, Luer lock to Luer slip
  • Environmentally Preferable:
  • Cat. No.:
    10847-610
  • Supplier no.:
    H93813701
Specifications
About this item
RAPIDFILL Connectors, Luer lock to Luer slip Connects with female Luer lock and Luer slip connections. Large finger grips protect against contamination.
  • Sterile Package
  • Contains DEHP
  • Not Made with Natural Rubber Latex
  • Blue color