Specifications
- Description:GloMax Explorer with Luminescence and Fluorescence, 1 each
- Size:1 each
- Environmentally Preferable:
- Cat. No.:PAGM3510
- Storage temperature:15...30 °C
- Supplier no.:GM3510
Specifications
About this item
The GloMax Explorer System is a high-performance multimode detection instrument developed with Promega reagents to provide a simple means of detecting advanced chemistries.
- Multimode Reader with Maximum Flexibility
- Fully loaded model detects luminescence, fluorescence and absorbance
- Integrated tablet PC provides quick and easy navigation
- Developed and optimized with our cell and gene reporter assays
The GloMax Explorer System is a high-performance multimode detection instrument developed with Promega reagents to provide a simple means of detecting advanced chemistries. This instrument provides researchers superior luminescence sensitivity and dynamic range for both strong and weak bioluminescence experimental samples as well as seamless integration with Promega industry-leading bioluminescence assays. GloMax Explorer provides flexible use of filters for fluorescence intensity, filtered luminescence and visible absorbance measurements. The instrument is operated by an integrated tablet PC, which provides quick and easy navigation through the control options. Exporting your results is easy with a variety of options, including export to your local data network. The GloMax Explorer software provides many of the required technical elements of a part 11-compliant system (user authentication and authorization, data integrity and protection, electronic signatures and audit trails) when used with the appropriate laboratory workflow.