ContactUsLinkComponent
GloMax 20/20 Luminometer, Promega
GloMax 20/20 Luminometer, Promega
Catalog # PAE5311
Supplier:  Avantor
undefined
GloMax 20/20 Luminometer, Promega
Catalog # PAE5311
Supplier:  Avantor
Specifications
  • Description:
    GloMax 20/20 Luminometer, 1 each
  • Size:
    1 each
  • Environmentally Preferable:
  • Cat. No.:
    PAE5311
  • Storage temperature:
    15...30 °C
  • Supplier no.:
    E5311
Specifications
About this item
The GloMax 20/20 Luminometer is an ultrasensitive, versatile and affordable luminometer designed for use with any Promega bioluminescent assay.
  • Ultrasensitive Single Tube Luminometer
  • Two fluorescence modules available
  • Optional single or dual auto-injectors
  • Ideal for use with our bioluminescent cell health and gene reporter assays
The GloMax 20/20 Luminometer combines instrumentation and software in a complete solution that includes bioluminescent assays, protocols and support. The GloMax 20/20 Luminometer is an ultrasensitive, versatile and affordable luminometer designed for use with any Promega bioluminescent assay. The touch screen interface provides comprehensive instrument control and data collection. Optional modules for fluorescence detection provide additional flexibility. The option of an intelligently designed internal auto-injection system is an added convenience and meets the demands of the Dual-Luciferase Assay. Software setup wizards guide the user through a brief process when establishing new protocols. New users can set up protocols and operate the instrument without a steep learning curve. Promega protocols are preloaded in the software to help users get started. The user can quickly select the protocol of interest and begin running assays directly to an Excel spreadsheet, where data can be analyzed quickly and easily. To learn more about the GloMax Systems, please visit: www.promega.com/products/instruments/