To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:STI1
- Clonality:Monoclonal
- Clone:1000
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10963
- Antigen Synonyms:Hsp70/Hsp90-organizing protein|STI1P60|Renal carcinoma antigen NY-REN-11|Hop|IEF-SSP-3521|Transformation-sensitive protein IEF SSP 3521|NY-REN-11 antigen|Hsc70/Hsp90-organizing protein|HOPSTI1L|stress-induced-phosphoprotein 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:STIP1 (NP_006810, 445 a.a. ~ 544 a.a) partial recombinant protein with GST tag.TKAMDVYQKALDLDSSCKEAADGYQRCMMAQYNRHDSPEDVKRRAMADPEVQQIMSDPAMRLILEQMQKDPQALSEHLKNPVIAQKIQKLMDVGLIAIR
- Purification:Protein A purified
- Cat. No.:103339-888
- Supplier no.:H00010963-M11
Specifications
About this item
The STI1 Antibody (1E3) from Novus Biologicals is a mouse monoclonal antibody to STI1. This antibody reacts with human, mouse, rat. The STI1 Antibody (1E3) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: STI1
Clonality: Monoclonal
Clone: 1000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat