To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sialic acid-binding Ig-like lectin 7
- Antigen Symbol:Siglec-7
- Clonality:Monoclonal
- Clone:3H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:27036
- Antigen Synonyms:sialic acid-binding Ig-like lectin 7|CD328|Siglec-7|p75/AIRM1|p75|sialic acid binding Ig-like lectin 7|adhesion inhibitory receptor molecule 1|CD328 antigen|siglec-7|AIRM1QA79 membrane protein|CDw328|sialic acid binding immunoglobulin-like lectin 7|D-siglec|Adhesion inhibitory receptor molecule 1|QA79|AIRM-1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SIGLEC7 (AAH28150, 1 a.a. ~ 468 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MLLLLLLPLLWGRERVEGQKSNRKDYSLTMQSSVTVQEGMCVHVRCSFSYPVDSQTDSDPVHGYWFRAGNDISWKAPVATNNPAWAVQEETRDRFHLLGDPQTKNCTLSIRDARMSDAGRYFFRMEKGNIKWNYKYDQLSVNVTALTHRPNILIPGTLESGCFQNLTCSVPWACEQGTPPMISWMGTSVSPLHPSTTRSSVLTLIPQPQHHGTSLTCQVTLPGAGVTTNRTIQLNVSYPPQNLTVTVFQGEGTAS
- Purification:IgG purified
- Cat. No.:103342-504
- Supplier no.:H00027036-M01
Specifications
About this item
The Siglec-7 / CD328 Antibody (3H1) from Novus Biologicals is a mouse monoclonal antibody to Siglec-7 / CD328. This antibody reacts with human. The Siglec-7 / CD328 Antibody (3H1) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Siglec-7
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human