To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:SF3B2
- Clonality:Monoclonal
- Clone:5D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10992
- Antigen Synonyms:Spliceosome-associated protein 145|SF3b1|Cus1|subunit 2|SAP145SAP 145|splicing factor 3b|145kDa|145kD|spliceosome associated protein 145|pre-mRNA splicing factor SF3b 145 kDa subunit|SF3b150|Pre-mRNA-splicing factor SF3b 145 kDa subunit|SF3b145|splicing factor 3B subunit 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SF3B2 (NP_006833, 592 a.a. ~ 646 a.a) partial recombinant protein with GST tag.YEGKEFETRLKEKKPGDLSDELRISLGMPVGPNAHKVPPPWLIAMQRYGPPPSY
- Purification:IgG purified
- Cat. No.:103339-924
- Supplier no.:H00010992-M01
Specifications
About this item
The SF3B2 Antibody (5D2) from Novus Biologicals is a mouse monoclonal antibody to SF3B2. This antibody reacts with human, mouse, rat. The SF3B2 Antibody (5D2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SF3B2
Clonality: Monoclonal
Clone: 5D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat