Anti-RPS8 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The RPS8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RPS8. This antibody reacts with human, zebrafish. The RPS8 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: RPS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Zebrafish
Specifications
- Size:100 μl
- Clonality:Polyclonal
- Purification:Immunogen affinity purified
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol
- Isotype:IgG
- Host:Rabbit
- Concentration:0.64 mg/ml
- Cat. No.:102711-328
- Reactivity:Zebrafish|Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:RPS8
- Gene ID:6202
- Immunogen:Recombinant protein encompassing a sequence within the center region of human RPS8. The exact sequence is proprietary.
- Conjugation:Unconjugated
- Antigen Synonyms:ribosomal protein S8,40S ribosomal protein S8|OK/SW-cl.83
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



