Order Entry
United States
ContactUsLinkComponent
Anti-PI16 Mouse Polyclonal Antibody
Anti-PI16 Mouse Polyclonal Antibody
  103347-830
Anti-PI16 Mouse Polyclonal Antibody
  103347-830
 :  H00221476-B01P
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Peptidase Inhibitor 16
  • Antigen Symbol:
    PI16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Gene ID:
    221476
  • Antigen Synonyms:
    PSP94-binding protein|PSPBP|MGC45378|protease inhibitor 16|PI-16|CRISP-9|dJ90K10.5|microseminoprotein|CRISP9|beta-binding protein|DKFZp586B1817|peptidase inhibitor 16|Cysteine-rich secretory protein 9|MSMBBP
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    PI16 (AAH22399.2, 1 a.a. ~ 408 a.a) full-length human protein.MHGSCSFLMLLLPLLLLLVATTGPVGALTDEEKRLMVELHNLYRAQVSPTASDMLHMRWDEELAAFAKAYARQCVWGHNKERGRRGENLFAITDEGMDVPLAMEEWHHEREHYNLSAATCSPGQMCGHYTQVVWAKTERIGCGSHFCEKLQGVEETNIELLVCNYEPPGNVKGKRPYQEGTPCSQCPSGYHCKNSLCEPIGSPEDAQDLPYLVTEAPSFRATEASDSRKMGTPSSLATGIPAFLVTEVSGSLATK
  • Purification:
    Protein A purified
  • Cat. No.:
    103347-830
  • Supplier no.:
    H00221476-B01P

 

 

The Peptidase Inhibitor 16 / PI16 Antibody from Novus Biologicals is a mouse polyclonal antibody to Peptidase Inhibitor 16 / PI16. This antibody reacts with human. The Peptidase Inhibitor 16 / PI16 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PI16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human