To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:PCAF
- Clonality:Monoclonal
- Clone:5E5.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8850
- Antigen Synonyms:P|EC 2.3.1.48|GCN5L|K(lysine) acetyltransferase 2B|Histone acetyltransferase PCAF|CREBBP-associated factor|P300/CBP-associated factorCAF|histone acetyltransferase KAT2B|Lysine acetyltransferase 2B|P/CAFGCN5|Histone acetylase PCAF|PCAF
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PCAF (AAH60823 353 a.a. - 452 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LEEEVYSQNSPIWDQDFLSASSRTSQLGIQTVINPPPVAGTISYNSTSSSLEQPNAGSSSPACKASSGLEANPGEKRKMTDSHVLEEAKKPRVMGDIPME
- Purification:IgG purified
- Cat. No.:103335-778
- Supplier no.:H00008850-M06
Specifications
About this item
The PCAF Antibody (5E5.) from Novus Biologicals is a mouse monoclonal antibody to PCAF. This antibody reacts with human, mouse. The PCAF Antibody (5E5.) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: PCAF
Clonality: Monoclonal
Clone: 5E5.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse