To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Oviductal glycoprotein 1, 120kDa
- Antigen Symbol:OVGP1
- Clonality:Monoclonal
- Clone:3F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Cross Adsorption:No
- Gene ID:5016
- Antigen Synonyms:120kDa|Oviductal glycoprotein|oviductin|OGP|oviductal glycoprotein 1|oviduct glycoprotein|mucin-9|oviduct-specific glycoprotein|Estrogen-dependent oviduct protein|mucin 9|EGP|MUC9|CHIT5
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:OVGP1 (NP_002548.3 579 a.a. - 677 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RNISVTPEGQTMPLRGENLTSEVGTHPRMGNLGLQMEAENRMMLSSSPVIQLPEQTPLAFDNRFVPIYGNHSSVNSVTPQTSPLSLKKEIPENSAVDEE
- Purification:IgG purified
- Cat. No.:103332-140
- Supplier no.:H00005016-M02
Specifications
About this item
The OVGP1 Antibody (3F9) from Novus Biologicals is a mouse monoclonal antibody to OVGP1. This antibody reacts with human. The OVGP1 Antibody (3F9) has been validated for the following applications: Western Blot, ELISA.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: OVGP1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human