To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Nesfatin-1/Nucleobindin-2
- Antigen Symbol:NUCB2
- Clonality:Monoclonal
- Clone:2062A
- Conjugation:DyLight® 550
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4925
- Antigen Synonyms:DNA-binding protein NEFA|nucleobinding 2|nucleobindin-2|NEFAGastric cancer antigen Zg4|nucleobindin 2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human NUCB2/Nesfatin-1 protein (amino acids 25-106) [UniProt P80303]
- Purification:Protein A or G purified
- Cat. No.:103294-860
- Supplier no.:MAB59491R
Specifications
About this item
The Nesfatin-1 / Nucleobindin-2 Antibody (2062A) [DyLight 550] from Novus Biologicals is a rabbit monoclonal antibody to Nesfatin-1 / Nucleobindin-2. This antibody reacts with human. The Nesfatin-1 / Nucleobindin-2 Antibody (2062A) [DyLight 550] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: NUCB2
Clonality: Monoclonal
Clone: 2062A
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human