To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:NEK8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:284086
- Antigen Synonyms:JCKNIMA-related kinase 12a|serine/thrionine-protein kinase NEK8|NEK12A|EC 2.7.11.1|NPHP9NIMA-family kinase NEK8|Nima-related protein kinase 12a|serine/threonine-protein kinase Nek8|EC 2.7.11|MGC138445|Never in mitosis A-related kinase 8|NIMA (never in mitosis gene a)- related kinase 8|nephrocystin 9|NimA-related protein kinase 8
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NEK8 (NP_835464.1, 1 a.a. ~ 692 a.a) full-length human protein.MEKYERIRVVGRGAFGIVHLCLRKADQKLVIIKQIPVEQMTKEERQAAQNECQVLKLLNHPNVIEYYENFLEDKALMIAMEYAPGGTLAEFIQKRCNSLLEEETILHFFVQILLALHHVHTHLILHRDLKTQNILLDKHRMVVKIGDFGISKILSSKSKAYTVVGTPCYISPELCEGKPYNQKSDIWALGCVLYELASLKRAFEAANLPALVLKIMSGTFAPISDRYSPELRQLVLSLLSLEPAQRPPLSHIMAQ
- Purification:Protein A purified
- Cat. No.:103348-116
- Supplier no.:H00284086-B01P
The NEK8 Antibody from Novus Biologicals is a mouse polyclonal antibody to NEK8. This antibody reacts with human. The NEK8 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: NEK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human