Anti-CTEN Rabbit Polyclonal Antibody
100 μg
Catalog # 103345-502
Marketsource
Supplier: Novus Biologicals
Supplier #: H00084951-D01P
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Tensin 4 / CTEN Antibody from Novus Biologicals is a rabbit polyclonal antibody to Tensin 4 / CTEN. This antibody reacts with human. The Tensin 4 / CTEN Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CTEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:100 μg
- Clonality:Polyclonal
- Purification:Protein A purified
- Storage Buffer:PBS (pH 7.4)
- Isotype:IgG
- Host:Rabbit
- Cat. No.:103345-502
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:CTEN
- Gene ID:84951
- Immunogen:TNS4 (AAH18706.1, 1 a.a. ~ 480 a.a) full-length human protein.MGSKASSPHGLGSPLVASPRLEKRLGGLAPQRGSRISVLSASPVSDVSYMFGSSQSLLHSSNSSHQSSSRSLESPANSSSSLHSLGSVSLCTRPSDFQAPRNPTLTMGQPRTPHSPPLAKEHASSCPPSITNSMVDIPIVLINGCPEPGSSPPQRTPGHQNSVQPGAASPSNPCPATRSNSQTLSDAPFTTCPEGPARDMQPTMKFVMDTSKYWFKPNITREQAIELLRKEEPGAFVIRDSSSYRGSFGLALKVQ
- Conjugation:Unconjugated
- Antigen Synonyms:tensin-4|C-terminal tensin-like protein|CTENC terminal tensin like|tensin 4|FLJ14950
- Antigen Name:Tensin 4
- ELISA:Yes
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



