To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:NCOA4
- Clonality:Monoclonal
- Clone:1F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 lambda
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8031
- Antigen Synonyms:PTC3RET-activating gene ELE1|ELE1NCoA-4|ARA70RFGret fused|Androgen receptor coactivator 70 kDa protein|nuclear receptor coactivator 4,70 kDa AR-activator|Androgen receptor-associated protein of 70 kDa|Ret-activating protein ELE1,70 kDa androgen receptor coactivator|DKFZp762E1112
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NCOA4 (NP_005428 505 a.a. - 614 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EGSPKEVPGTEDRAGKQKFKSPMNTSWCSFNTADWVLPGKKMGNLSQLSSGEDKWLLRKKAQEVLLNSPLQEEHNFPPDHYGLPAVCDLFACMQLKVDKEKWLYRTPLQM
- Purification:IgG purified
- Cat. No.:103334-616
- Supplier no.:H00008031-M05
Specifications
About this item
The NCOA4 Antibody (1F11) from Novus Biologicals is a mouse monoclonal antibody to NCOA4. This antibody reacts with human. The NCOA4 Antibody (1F11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NCOA4
Clonality: Monoclonal
Clone: 1F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human