To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:MPG
- Clonality:Monoclonal
- Clone:1G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:4350
- Antigen Synonyms:MPG|ADPG|localized 68 kb upstream the humanzeta globin gene on16p|3' end of the Mid1 gene|CRA36.1|3-methyladenine DNA glycosidase|EC 3.2.2.21|N-methylpurine-DNA glycosylasePIG16|AAG|proliferation-inducing protein 16|Mid1|PIG11|N-methylpurine-DNA glycosylase|3-alkyladenine DNA glycosylase|DNA-3-methyladenine glycosylase|MDGalkyladenine DNA glycosylase|proliferation-inducing protein 11|APNG|CRA36.1 (3-methyl-adenine DNA glycosylase)|ANPG
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MPG (AAH14991, 1 a.a. - 90 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPARSGAQFCRRMGQKKQRPARAGQPHSSSDAAQAPAEQPHSSSDAAQAPCPRERCLGPPTTPGPYRSIYFSSPKGHLTRLGLEFFDQPA
- Purification:IgG purified
- Cat. No.:103331-018
- Supplier no.:H00004350-M10
The MPG Antibody (1G6) from Novus Biologicals is a mouse monoclonal antibody to MPG. This antibody reacts with human. The MPG Antibody (1G6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: MPG
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human