To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MAT1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4143
- Antigen Synonyms:MAT-I/III|alpha|MATA1MAT 1|methionine adenosyltransferase I|S-adenosylmethionine synthetase isoform type-1|adoMet synthetase 1|Methionine adenosyltransferase 1|SAMS1EC 2.5.1.6|AMS1|SAMS|AdoMet synthase 1|S-adenosylmethionine synthase isoform type-1|Methionine adenosyltransferase I/III|MAT
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against a recombinant protein corresponding to amino acids: MNGPVDGLCDHSLSEGVFMFTSESV
- Purification:Immunogen affinity purified
- Cat. No.:103283-064
- Supplier no.:NBP2-33533
Specifications
About this item
The MAT1A Antibody from Novus Biologicals is a rabbit polyclonal antibody to MAT1A. This antibody reacts with human. The MAT1A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: MAT1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human