To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:MAPKAP Kinase 3
- Antigen Symbol:MAPKAPK3
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:7867
- Antigen Synonyms:mitogen-activated protein kinase-activated protein kinase 3|MAPKAP33pK|3PKEC 2.7.11.1|MAPKAP kinase 3|EC 2.7.11|Chromosome 3p kinase|MAPKAPK-3|MAP kinase-activated protein kinase 3|MAPK-activated protein kinase 3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MAPKAPK3 (NP_004626, 59 a.a. - 153 a.a.) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LECFHRRTGQKCALKLLYDSPKARQEVDHHWQASGGPHIVCILDVYENMHHGKRCLLIIMECMEGGELFSRIQERGDQAFTEREAAEIMRDIGTA
- Purification:IgG purified
- Cat. No.:103334-492
- Supplier no.:H00007867-M04
The MAPKAP Kinase 3 Antibody (1C3) from Novus Biologicals is a mouse monoclonal antibody to MAPKAP Kinase 3. This antibody reacts with human. The MAPKAP Kinase 3 Antibody (1C3) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MAPKAPK3
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human