To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:IL1F8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:27177
- Antigen Synonyms:IL1F8 (Canonical product IL-1F8a)|FIL1 eta|FIL1|FIL1H|family of interleukin 1-eta|Interleukin-1 eta|interleukin 1 family|IL-1F8IL1-ETA|IL-1H2MGC126880|member 8 (eta)|interleukin-1 family member 8|IL1H2MGC126882|Interleukin-1 Superfamily e|Interleukin-1 homolog 2|interleukin 1|IL-1 eta|eta|FIL1-(ETA)|IL-1F8 (FIL1-eta)
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:PKSYAIRDSRQMVWVLSGNSLIAAPLSRSIKPVTLHLIACRDTEFSDKEKGNMVYLGIKGKDLCLFCAEIQGKP
- Purification:Immunogen affinity purified
- Cat. No.:103266-502
- Supplier no.:NBP1-83892
The IL1F8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to IL1F8. This antibody reacts with human. The IL1F8 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: IL1F8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human