To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GUCY2D
- Clonality:Monoclonal
- Clone:4E12.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3000
- Antigen Synonyms:GUC1A4|EC 4.6.1|Rod outer segment membrane guanylate cyclase|retGC|ROS-GC1|LCA1RCD2|guanylate cyclase 2D|Guanylate cyclase 2D|retinal|membrane (retina-specific)|RETGC1|GUC2DCORD5|ROS-GC|LCA|retinal guanylyl cyclase 1|CYGD|RETGC-1EC 4.6.1.2|ROSGC|CORD6cone rod dystrophy 5/6
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GUCY2D (NP_000171, 521 a.a. - 630 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RKVAQGSRSSLGARSMSDIRSGPSQHLDSPNIGVYEGDRVWLKKFPGDQHIAIRPATKTAFSKLQELRHENVALYLGLFLARGAEGPAALWEGNLAVVSEHCTRGSLQDL
- Purification:IgG purified
- Cat. No.:103327-796
- Supplier no.:H00003000-M01
Specifications
About this item
The GUCY2D Antibody (4E12.) from Novus Biologicals is a mouse monoclonal antibody to GUCY2D. This antibody reacts with human. The GUCY2D Antibody (4E12.) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: GUCY2D
Clonality: Monoclonal
Clone: 4E12.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human