To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:GPR89
- Clonality:Monoclonal
- Clone:4D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:51463
- Antigen Synonyms:GPHR|Protein GPR89B|GPR89|GPHRB|Golgi pH regulator A|G protein-coupled receptor 89|protein GPR89|putative MAPK-activating protein PM01|Golgi pH regulator B|SH120|FLJ55546|putative NF-kappa-B-activating protein 90|G protein-coupled receptor 89B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GPR89 (AAH03187, 175 a.a. ~ 285 a.a) partial recombinant protein with GST tag.SYFLRNVTDTDILALERRLLQTMDMIISKKKRMAMARRTMFQKGEVHNKPSGFWGMIKSVTTSASGSENLTLIQQEVDALEELSRQLFLETADLYATKERIEYSKTFKGK
- Purification:IgG purified
- Cat. No.:103344-184
- Supplier no.:H00051463-M01
The GPR89 Antibody (4D8) from Novus Biologicals is a mouse monoclonal antibody to GPR89. This antibody reacts with human. The GPR89 Antibody (4D8) has been validated for the following applications: ELISA.
Type: Primary
Antigen: GPR89
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human