To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:GOLPH4
- Clonality:Monoclonal
- Clone:5E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:27333
- Antigen Synonyms:Golgi phosphoprotein 4130 kDa golgi-localized phosphoprotein|Golgi phosphoprotein of 130 kDa|golgi integral membrane protein 4|GIMPc|GIMPCcis|GPP130Golgi-localized phosphoprotein of 130 kDa
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GOLPH4 (NP_055313, 473 a.a. ~ 569 a.a) partial recombinant protein with GST tag.HQEQLRQQAHYDAMDNDIVQGAEDQGIQGEEGAYERDNQHQDEAEGDPGNRHEPREQGPREADPESEADRAAVEDINPADDPNNQGEDEFEEAEQV
- Purification:IgG purified
- Cat. No.:103342-810
- Supplier no.:H00027333-M01
The GOLPH4 Antibody (5E11) from Novus Biologicals is a mouse monoclonal antibody to GOLPH4. This antibody reacts with human. The GOLPH4 Antibody (5E11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: GOLPH4
Clonality: Monoclonal
Clone: 5E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human