To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Guanine nucleotide binding protein (G protein), Q polypeptide
- Antigen Symbol:GNAQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Gene ID:2776
- Antigen Synonyms:q polypeptide|guanine nucleotide binding protein (G protein)|guanine nucleotide-binding protein G(q) subunit alpha|GAQG-ALPHA-q|Guanine nucleotide-binding protein alpha-q
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GNAQ (AAH57777, 1 a.a. - 359 a.a.) full-length human protein. MTLESIMACCLSEEAKEARRINDEIERQLRRDKRDARRELKLLLLGTGESGKSTFIKQMRIIHGSGYSDEDKRGFTKLVYQNIFTAMQAMIRAMDTLKIPYKYEHNKAHAQLVREVDVEKVSAFENPYVDAIKSLWNDPGIQECYDRRREYQLSDSTKYYLNDLDRVADPAYLPTQQDVLRVRVPTTGIIEYPFDLQSVIFRMVDVGGQRSERRKWIHCFENVTSIMFLVALSEYDQVLVESDNENRMEESKALFRTIITYPWFQNSSVILFLNKKDLLEEKIMYSHLVDYFPEYDGPQRDAQAAREFILKMFVDLNPDSDKIIYSHFTCATDTENIRFVFAAVKDTILQLNLKEYNLV
- Purification:Protein G purified
- Cat. No.:103327-336
- Supplier no.:H00002776-B01P
The GNAQ Antibody from Novus Biologicals is a mouse polyclonal antibody to GNAQ. This antibody reacts with human, rat. The GNAQ Antibody has been validated for the following applications: Western Blot.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: GNAQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat