To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GM-CSF R alpha
- Clonality:Monoclonal
- Clone:2G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1438
- Antigen Synonyms:CD116 antigen|alpha|GM-CSF receptor alpha subunit|MGC4838|GMCSFR|colony stimulating factor 2 receptor alpha subunit|GMCSFR-alpha|granulocyte-macrophage colony-stimulating factor receptor subunit alpha|GMR-alpha|CSF2RAX|CSF2RY|CSF2RAY|GMR|colony stimulating factor 2 receptor|granulocyte-macrophage colony-stimulating factor receptor alpha chain|GM-CSF-R-alpha|CDw116|MGC3848|low-affinity(granulocyte-macrophage)|CSF2R|CD116|CSF2RX
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CSF2RA (AAH02635.1, 1 a.a. - 400 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLLLVTSLLLCELPHPAFLLIPEKSDLRTVAPASSLNVRFDSRTMNLSWDCQENTTFSKCFLTDKKNRVVEPRLSNNECSCTFREICLHEGVTFEVHVNTSQRGFQQKLLYPNSGREGTAAQNFSCFIYNADLMNCTWARGPTAPRDVQYFLYIRNSKRRREIRCPYYIQDSGTHVGCHLDNLSGLTSRNYFLVNGTSREIGIQFFDSLLDTKKIERFNPPSNVTVRCNTTHCLVRWKQPRTYQKLSYLDFQYQLDVHRKNTQPGTENLLINVSGDLENRYNFPSSEPRAKHSVKIRAADVRILNWSSWSEAIEFGSDDGNLGSVYIYVLLIVGTLVCGIVLGFLFKRFLRIQRLFPPVPQIKDKLNDNHEVEDEIIWEEFTPEEGKGYREEVLTVKEIT
- Tested Applications:Proximity Ligation Assay
- Purification:IgG purified
- Cat. No.:103324-476
- Supplier no.:H00001438-M03
Specifications
About this item
The GM-CSF R alpha Antibody (2G5) from Novus Biologicals is a mouse monoclonal antibody to GM-CSF R alpha. This antibody reacts with human. The GM-CSF R alpha Antibody (2G5) has been validated for the following applications: Western Blot, ELISA, Proximity Ligation Assay.
Type: Primary
Antigen: GM-CSF R alpha
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human