Order Entry
United States
ContactUsLinkComponent
 
Anti-Ferritin Light Chain Mouse Monoclonal Antibody [clone: X3]
  103326-806
 :  Avantor
Anti-Ferritin Light Chain Mouse Monoclonal Antibody [clone: X3]
  103326-806
 :  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Ferritin Light Chain
  • Clonality:
    Monoclonal
  • Clone:
    X3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    2512
  • Antigen Synonyms:
    ferritin|Ferritin L subunit|ferritin light chain|ferritin L-chain|NBIA3|light polypeptide|MGC71996|ferritin light polypeptide-like 3
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    FTL (AAH04245, 1 a.a. - 175 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSSQIRQNYSTDVEAAVNSLVNLYLQASYTYLSLGFYFDRDDVALEGVSHFFRELAEEKREGYERLLKMQNQRGGRALFQDIKKPAEDEWGKTPDAMKAAMALEKKLNQALLDLHALGSARTDPHLCDFLETHFLDEEVKLIKKMGDHLTNLHRLGGPEAGLGEYLFERLTLKHD
  • Purification:
    IgG purified
  • Cat. No.:
    103326-806
  • Supplier no.:
    H00002512-M18

 

 

The Ferritin Light Chain Antibody (X3) from Novus Biologicals is a mouse monoclonal antibody to Ferritin Light Chain. This antibody reacts with human. The Ferritin Light Chain Antibody (X3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, RNA Inhibition.

Type: Primary
Antigen: Ferritin Light Chain
Clonality: Monoclonal
Clone: X3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human