To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome b5 type A
- Clonality:Monoclonal
- Clone:4C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1528
- Antigen Synonyms:MCB5cytochrome b5|Microsomal cytochrome b5 type A|type 1 cyt-b5|cytochrome b5 type A (microsomal)|cytochrome b5 (microsomal)|cytochrome b-5|CYB5
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CYB5A (AAH15182, 1 a.a. - 134 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAEQSDEAVKYYTLEEIQKHNHSKSTWLILHHKVYDLTKFLEEHPGGEEVLREQAGGDATENFEDVGHSTDAREMSKTFIIGELHPDDRPKLNKPPETLITTIDSSSSWWTNWVIPAISAVAVALMYRLYMAED
- Purification:IgG purified
- Cat. No.:103324-716
- Supplier no.:H00001528-M05
Specifications
About this item
The Cytochrome b5 type A Antibody (4C2) from Novus Biologicals is a mouse monoclonal antibody to Cytochrome b5 type A. This antibody reacts with human. The Cytochrome b5 type A Antibody (4C2) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Cytochrome b5 type A
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human