To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Collagen VII
- Clonality:Monoclonal
- Clone:LH7.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1294
- Antigen Synonyms:epidermolysis bullosa|EBD1|collagen VII|alpha 1|EBR1|long chain collagen|type VII|Collagen-7|dominant and recessive|collagen|dystrophic|collagen alpha-1(VII) chain|EBDCT|alpha-1 polypeptide|Long-chain collagen|LC collagen|Collagen 7
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed with Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions.
- Purification:Tissue culture supernatant
- Cat. No.:103357-458
Specifications
About this item
The Collagen VII Antibody (LH7.2) from Novus Biologicals is a mouse monoclonal antibody to Collagen VII. This antibody reacts with human. The Collagen VII Antibody (LH7.2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Collagen VII
Clonality: Monoclonal
Clone: LH7.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human