To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:FBXO22
- Clonality:Monoclonal
- Clone:6G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:26263
- Antigen Synonyms:F-box only protein 22|MGC31799|FIST domain containing 1|FBX22FLJ13986|F-box protein FBX22p44|F-box protein 22|FISTC1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FBXO22 (AAH39024, 1 a.a. ~ 300 a.a) full length recombinant protein with GST tag.MADSETFISLEECRGHKRARKRTSMETALALEKLFPKQCQVLGIVTPGIVVTPMGSGSNRPQEIEIGESGFALLFPQIEGIKIQPFHFIKDPKNLTLERHQLTEVGLLDNPELRVVLVFGYNCCKVGASNYLQQVVSTFSDMNIILAGGQVDNLSSLTSEKNPLDIDASGVVGLSFSGHRIQSATVLLNEDVSDEKTAEAAMQRLKAANIPEHNTIGFMFACVGRGFQYYRAKGNVEADAFRKFFPSVPLFGFFG
- Purification:IgG purified
- Cat. No.:103342-250
- Supplier no.:H00026263-M01
The FBXO22 Antibody (6G9) from Novus Biologicals is a mouse monoclonal antibody to FBXO22. This antibody reacts with human. The FBXO22 Antibody (6G9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: FBXO22
Clonality: Monoclonal
Clone: 6G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human