To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Connexin 30.3
- Antigen Symbol:Cx30.3
- Clonality:Monoclonal
- Clone:1E3-1C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:127534
- Antigen Synonyms:30.3kDa|beta 4|MGC21116|beta 4 (connexin 30.3)|gap junction beta-4 protein|Cx30.3|gap junction protein|EKV|connexin-30.3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GJB4 (AAH34709, 1 a.a. ~ 267 a.a) full length recombinant protein with GST tag.MNWAFLQGLLSGVNKYSTVLSRIWLSVVFIFRVLVYVVAAEEVWDDEQKDFVCNTKQPGCPNVCYDEFFPVSHVRLWALQLILVTCPSLLVVMHVAYREERERKHHLKHGPNAPSLYDNLSKKRGGLWWTYLLSLIFKAAVDAGFLYIFHRLYKDYDMPRVVACSVEPCPHTVDCYISRPTEKKVFTYFMVTTAAICILLNLSEVFYLVGKRCMEIFGPRHRRPRCRECLPDTCPPYVLSQGGHPEDGNSVLMKA
- Purification:IgG purified
- Cat. No.:103346-690
- Supplier no.:H00127534-M01
The Connexin 30.3 / GJB4 Antibody (1E3-1C12) from Novus Biologicals is a mouse monoclonal antibody to Connexin 30.3 / GJB4. This antibody reacts with human. The Connexin 30.3 / GJB4 Antibody (1E3-1C12) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Cx30.3
Clonality: Monoclonal
Clone: 1E3-1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human