To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CUGBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10659
- Antigen Synonyms:NAPOR-2|CELF-2|CUGBP Elav-like family member 2|RNA-binding protein 2|RNA binding protein 2|CUG-BP- and ETR-3-like factor 2|CUGBP|Elav-like family member 2|ETR3RNA-binding protein BRUNOL-3|Bruno-like protein 3|CUG triplet repeat|Etr-3|CUG triplet repeat RNA-binding protein 2|CUGBP2hNAPOR|Neuroblastoma apoptosis-related RNA-binding protein|BRUNOL3ELAV-type RNA-binding protein 3|CUG-BP2|NAPOR
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:CUGBP2 (AAH36391.1, 1 a.a. ~ 483 a.a) full-length human protein.MMVEGRLLVPDRINGTANKMNGALDHSDQPDPDAIKMFVGQIPRSWSEKELKELFEPYGAVYQINVLRDRSQNPPQSKGCCFVTFYTRKAALEAQNALHNIKTLPGMHHPIQMKPADSEKSNAVEDRKLFIGMVSKKCNENDIRVMFSPFGQIEECRILRGPDGLSRGCAFVTFSTRAMAQNAIKAMHQSQTMEGCSSPIVVKFADTQKDKEQRRLQQQLAQQMQQLNTATWGNLTGLGGLTPQYLALLQQATSS
- Purification:Protein A purified
- Cat. No.:103339-374
- Supplier no.:H00010659-D01P
Specifications
About this item
The CUGBP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CUGBP2. This antibody reacts with human, mouse. The CUGBP2 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CUGBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse