Order Entry
United States
ContactUsLinkComponent
 
Anti-CMA1 Goat Polyclonal Antibody
Catalog # 103408-410
Supplier:  Novus Biologicals
Anti-CMA1 Goat Polyclonal Antibody
Catalog # 103408-410
Supplier:  Novus Biologicals
Supplier Number:  NBP2-27551
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chymase
  • Antigen Symbol:
    CMA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    1215
  • Antigen Synonyms:
    MGC119890|heart|mast cell|EC 3.4.21|EC 3.4.21.39|CYH|chymase|chymase 1 preproprotein transcript E|MCT1|chymase 1|chymase 1 preproprotein transcript I|MGC119891|Alpha-chymase|Mast cell protease I|CYM
  • Storage Buffer:
    0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Peptide with sequence C-QFRHPKYNTSTLHHD corresponding to internal region according to NP_001827.1.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103408-410

Specifications

About this item

The Chymase / CMA1 / Mast Cell Chymase Antibody from Novus Biologicals is a goat polyclonal antibody to Chymase / CMA1 / Mast Cell Chymase. This antibody reacts with human. The Chymase / CMA1 / Mast Cell Chymase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Type: Primary
Antigen: CMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human