Order Entry
United States
ContactUsLinkComponent
 
Anti-Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Mouse Monoclonal Antibody [clone: 3A9]
Catalog # 103402-878
Supplier:  Novus Biologicals
Anti-Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Mouse Monoclonal Antibody [clone: 3A9]
Catalog # 103402-878
Supplier:  Novus Biologicals
Supplier Number:  NBP2-45527
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2
  • Clonality:
    Monoclonal
  • Clone:
    3A9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    10678
  • Antigen Synonyms:
    B3GN-T1|beta-1,3-Gn-T1|BGNT2|BGnT-1|b3Gal-T7|B3GNT-2|Beta-1,3-galactosyltransferase 7|UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7|Beta-1,3-N-acetylglucosaminyltransferase 1|B3GALT7|beta-1,3-N-acetylglucosaminyltransferase bGnT-1|B3GNT1|Beta-1,3-Gn-T2|B3GN-T2|EC 2.4.1.-|Beta3GalT7|UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1|Beta-3-Gx-T7|beta3Gn-T2|BGnT-2|UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2|BETA3GNT|Beta-1,3-N-acetylglucosaminyltransferase 2|Beta3Gal-T7|UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7|Beta-1,3-GalTase 7|beta-1,3-N-acetylglucosaminyltransferase bGnT-2|Beta3Gn-T1|B3GNT
  • Storage Buffer:
    PBS (pH 7.3), 1.0% BSA and 50% Glycerol
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Full length human recombinant protein of human B3GNT2 (NP_006568) produced in HEK293T cell.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103402-878

Specifications

About this item

The Beta-1,3-N-Acetylglucosaminyltransferase 2 / B3GNT2 Antibody (3A9) from Novus Biologicals is a mouse monoclonal antibody to Beta-1,3-N-Acetylglucosaminyltransferase 2 / B3GNT2. This antibody reacts with human. The Beta-1,3-N-Acetylglucosaminyltransferase 2 / B3GNT2 Antibody (3A9) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunofluorescence.

Type: Primary
Antigen: Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2
Clonality: Monoclonal
Clone: 3A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human