To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Aspartate Aminotransferase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Epitope:C-RSYRYWDAEKR
- Gene ID:2805
- Antigen Synonyms:soluble (aspartate aminotransferase 1)|Transaminase A|cytoplasmic|glutamic-oxaloacetic transaminase 1|aspartate aminotransferase|GIG18|EC 2.6.1.1|growth-inhibiting protein 18|Glutamate oxaloacetate transaminase 1
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-RSYRYWDAEKR corresponding to internal region according to NP_002070.1.
- Purification:Immunogen affinity purified
- Cat. No.:103408-276
The Aspartate Aminotransferase Antibody from Novus Biologicals is a goat polyclonal antibody to Aspartate Aminotransferase. This antibody reacts with human. The Aspartate Aminotransferase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: Aspartate Aminotransferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-RSYRYWDAEKR
Host: Goat
Isotype: IgG
Reactivity: Human