To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:T-lymphocyte surface antigen Ly-9
- Antigen Symbol:LY9
- Clonality:Monoclonal
- Clone:MM0163-3L25
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4063
- Antigen Synonyms:mLY9|CD229 antigen|lymphocyte antigen 9Cell surface molecule Ly-9|cell-surface molecule Ly-9|hly9|T-lymphocyte surface antigen Ly-9|SLAMF3|CD229
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Human Lymphocyte Antigen 9
- Purification:>95% pure by SDS-PAGE
- Cat. No.:103400-214
Specifications
About this item
The CD229 / SLAMF3 / Lymphocyte Antigen 9 Antibody (MM0163-3L25) [Allophycocyanin] from Novus Biologicals is a mouse monoclonal antibody to CD229 / SLAMF3 / Lymphocyte Antigen 9. This antibody reacts with human. The CD229 / SLAMF3 / Lymphocyte Antigen 9 Antibody (MM0163-3L25) [Allophycocyanin] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: LY9
Clonality: Monoclonal
Clone: MM0163-3L25
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human