To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD1c/BDCA-1
- Clonality:Monoclonal
- Clone:M241
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:911
- Antigen Synonyms:BDCA1|CD1c antigen|CD1c molecule|CD1A|T-cell surface glycoprotein CD1c|CD1|differentiation antigen CD1-alpha-3|R7|c polypeptide|cortical thymocyte antigen CD1C
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:MOLT 4 cell line
- Purification:Protein A or G purified
- Cat. No.:103357-260
Specifications
About this item
The CD1c / BDCA-1 Antibody (M241) from Novus Biologicals is a mouse monoclonal antibody to CD1c / BDCA-1. This antibody reacts with human. The CD1c / BDCA-1 Antibody (M241) has been validated for the following applications: Immunohistochemistry, Immunoprecipitation, Radioimmunoassay.
Type: Primary
Antigen: CD1c/BDCA-1
Clonality: Monoclonal
Clone: M241
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human