Order Entry
United States
ContactUsLinkComponent
Anti-CCRL2/CRAM-A/B Rabbit Polyclonal Antibody
Anti-CCRL2/CRAM-A/B Rabbit Polyclonal Antibody
Catalog # 102706-632
Supplier:  Novus Biologicals
Anti-CCRL2/CRAM-A/B Rabbit Polyclonal Antibody
Catalog # 102706-632
Supplier:  Novus Biologicals
Supplier Number:  NBP2-15769
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CCRL2/CRAM-A/B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    9034
  • Antigen Synonyms:
    Chemokine receptor CCR11|Putative MCP-1 chemokine receptor|CRAM-B|CKRXMGC116710|CCR11|CRAM-A|HCRMGC34104|CCR6|C-C chemokine receptor-like 2|FLJ55815|Chemokine receptor X|CRAM|chemokine (C-C motif) receptor-like 2
  • Storage Buffer:
    PBS (pH 7.0), 1.0% BSA and 20% Glycerol
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CCRL2. The exact sequence is proprietary.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102706-632
  • Supplier no.:
    NBP2-15769

Specifications

About this item

The CCRL2 / CRAM-A / B Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCRL2 / CRAM-A / B. This antibody reacts with human, mouse. The CCRL2 / CRAM-A / B Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CCRL2/CRAM-A/B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse