To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Bcl-10
- Clonality:Monoclonal
- Clone:BL10/411
- Conjugation:DyLight® 405
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:8915
- Antigen Synonyms:CARD-containing apoptotic signaling protein|CED-3/ICH-1 prodomain homologous E10-like regulator|cCARMEN|Cellular homolog of vCARMEN|caspase-recruiting domain-containing protein|B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10|CARD-containing molecule enhancing NF-kappa-B|hCLAP|Mammalian CARD-containing adapter molecule E10|CARMEN|CARD-containing proapoptotic protein|CARD-like apoptotic protein|cellular-E10|c-E10bcl-10|mE10CARD containing molecule enhancing NF-kB|Bcl-10
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Human BCL10 recombinant protein (epitope aa122-168)
- Purification:Protein G purified
- Cat. No.:103404-180
Specifications
About this item
The Bcl-10 Antibody (BL10 / 411) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to Bcl-10. This antibody reacts with human. The Bcl-10 Antibody (BL10 / 411) [DyLight 405] has been validated for the following applications: ELISA.
Type: Primary
Antigen: Bcl-10
Clonality: Monoclonal
Clone: BL10/411
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human