To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:ATG9A
- Clonality:Monoclonal
- Clone:Atg9 14F2 8B1
- Conjugation:Biotin
- ELISA:Yes
- Host:Hamster
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:79065
- Antigen Synonyms:APG9 autophagy 9-like 1 (S. cerevisiae)|APG9 autophagy 9-like 1|autophagy-related protein 9A|ATG9 autophagy related 9 homolog A (S. cerevisiae)|MGD3208|APG9L1APG9-like 1|autophagy 9-like 1 protein|mATG9|FLJ22169
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Conserved C-terminal synthetic peptide of Human ATG9 sequence
- Purification:Protein G purified
- Cat. No.:103407-564
The ATG9A Antibody (Atg9 14F2 8B1) [Biotin] from Novus Biologicals is a hamster monoclonal antibody to ATG9A. This antibody reacts with human, mouse, rat. The ATG9A Antibody (Atg9 14F2 8B1) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
Type: Primary
Antigen: ATG9A
Clonality: Monoclonal
Clone: Atg9 14F2 8B1
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human, Mouse, Rat