To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ATG12
- Clonality:Monoclonal
- Clone:948
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:9140
- Antigen Synonyms:ubiquitin-like protein ATG12|Apg12 (autophagy 12|Autophagy-related protein 12|APG12-like|S. cerevisiae)-like|FBR93|yeast) homolog|APG12HAPG12|APG12 autophagy 12-like (S. cerevisiae)|ATG12 autophagy related 12 homolog (S. cerevisiae)
- Storage Buffer:PBS
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:The exact sequence is proprietary.
- Purification:Protein G purified
- Cat. No.:103403-768
Specifications
About this item
The ATG12 Antibody (948) from Novus Biologicals is a mouse monoclonal antibody to ATG12. This antibody reacts with human, mouse. The ATG12 Antibody (948) has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ATG12
Clonality: Monoclonal
Clone: 948
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse